Learn More
Medchemexpress LLC Raloxifene hydrochloride | 82640-04-8 | 99.8% | 510.04 | 25 MG
SDP
Catalog No. 5000351834
Encompass_Preferred
Raloxifene hydrochloride is a second-generation, selective, and orally active estrogen receptor modulator. It exhibits estrogen-agonistic effects on bone and lipid metabolism, while demonstrating estrogen-antagonistic effects on uterine endometrium and breast tissue.
  • Activates TGF beta 3 promoter as a full agonist
  • Inhibits estrogen response element-containing vitellogenin promoter expression
  • Acts as a potent uncompetitive inhibitor of human liver aldehyde oxidase
  • Functions as a noncompetitive inhibitor of aldehyde oxidase-catalyzed reduction reactions
  • Decreases viability in mouse mammary carcinoma cells
  • Demonstrates estrogenic activity on bone resorption and serum cholesterol in ovariectomized rats
  • Increases bone mineral density in ovariectomized rats
  • Reduces serum cholesterol in ovariectomized rats
  • Suppresses tumor volumes and decreases lymph node metastasis in mice
Catalog No. 50-003-51834 Supplier Medchemexpress LLC Supplier No. 25MGHY13738AR
Raloxifene hydrochloride is a second-generation, selective, and orally active estrogen receptor modulator. It exhibits estrogen-agonistic effects on bone and lipid metabolism, while demonstrating estrogen-antagonistic effects on uterine endometrium and breast tissue.
  • Activates TGF beta 3 promoter as a full agonist
  • Inhibits estrogen response element-containing vitellogenin promoter expression
  • Acts as a potent uncompetitive inhibitor of human liver aldehyde oxidase
  • Functions as a noncompetitive inhibitor of aldehyde oxidase-catalyzed reduction reactions
  • Decreases viability in mouse mammary carcinoma cells
  • Demonstrates estrogenic activity on bone resorption and serum cholesterol in ovariectomized rats
  • Increases bone mineral density in ovariectomized rats
  • Reduces serum cholesterol in ovariectomized rats
  • Suppresses tumor volumes and decreases lymph node metastasis in mice