Learn More
Medchemexpress LLC Raloxifene (Keoxifene) | 84449-90-1 | MFCD00866415 | 99.6% | 473.58 | 25 MG
SDP
Catalog No. 5000349779
Encompass_Preferred
Raloxifene (Keoxifene) is a benzothiophene-derived selective estrogen receptor modulator (SERM). It exhibits estrogen-agonistic effects on bone and lipids, and estrogen-antagonistic effects on the breast and uterus. Raloxifene is utilized in research for breast cancer and osteoporosis.
  • Benzothiophene-derived selective estrogen receptor modulator
  • Exhibits estrogen-agonistic effects on bone and lipids
  • Exhibits estrogen-antagonistic effects on breast and uterus
  • Utilized in research for breast cancer
  • Utilized in research for osteoporosis
Catalog No. 50-003-49779 Supplier Medchemexpress LLC Supplier No. 25MGHY13738
Raloxifene (Keoxifene) is a benzothiophene-derived selective estrogen receptor modulator (SERM). It exhibits estrogen-agonistic effects on bone and lipids, and estrogen-antagonistic effects on the breast and uterus. Raloxifene is utilized in research for breast cancer and osteoporosis.
  • Benzothiophene-derived selective estrogen receptor modulator
  • Exhibits estrogen-agonistic effects on bone and lipids
  • Exhibits estrogen-antagonistic effects on breast and uterus
  • Utilized in research for breast cancer
  • Utilized in research for osteoporosis