Learn More
ATCC Ralstonia pickettii
Catalog No. 502382945
Encompass
Ralstonia pickettii strain [ICPB 3981, K-288] was isolated from a patient who had undergone a tracheotomy. This culture is a whole-genome sequenced bacterial type strain.
Catalog No. 50-238-2945 Supplier ATCC Supplier No. 27511
Ralstonia pickettii strain [ICPB 3981, K-288] was isolated from a patient who had undergone a tracheotomy. This culture is a whole-genome sequenced bacterial type strain.