Learn More
Cayman Chemical Raltegravir-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2712343-38-7, 1 mg
SDP
Catalog No. 502936911
Encompass_Preferred
An internal standard for the quantification of raltegravir by GC- or LC-MS
Catalog No. 50-293-6911 Supplier Cayman Chemical Supplier No. 264581 MG
An internal standard for the quantification of raltegravir by GC- or LC-MS