Learn More
Apexbio Technology LLC Raltegravir (MK-0518), 10mg. Cas: 518048-05-0 MFCD: MFCD10698872. HIV-1 integrase inhibitor
SDP
Catalog No. 501904872
Encompass_Preferred
Raltegravir, formerly named MK-0518, is an HIV-1 integrase strand transfer inhibitor which has been shown to have activity against multidrug-resistant HIV-1 and both CCR5-trophic and CXCR4-trophic HIV-1 in vitro. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4872 Supplier Apexbio Technology LLC Supplier No. A407310
Add to Cart
Add to Cart
Raltegravir, formerly named MK-0518, is an HIV-1 integrase strand transfer inhibitor which has been shown to have activity against multidrug-resistant HIV-1 and both CCR5-trophic and CXCR4-trophic HIV-1 in vitro. Other sizes are available. Please inqury us for quote.