Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Raltegravir potassium | 871038-72-1 | MFCD12031642 | 99.8% | 482.51 | C20H20FKN6O5 | 25 MG
SDP

Catalog No. 5000249311
Encompass_Preferred

Raltegravir potassium is the potassium salt of raltegravir, a potent HIV integrase strand transfer inhibitor widely used in research to study viral integration and screening assays. It is supplied in solid and solution formats and is intended for laboratory research use only.

  • Potent HIV integrase inhibitor for research applications.
  • Potassium salt form to improve handling and solubility properties.
  • Molecular formula C20H20FKN6O5, molecular weight 482.51.
  • High purity (approximately 99.8%), suitable for analytical work.
  • Available in multiple formats including 25 mg solid and 10 mM in DMSO solutions.
  • Intended for in vitro and analytical research use only; not for human use.

Catalog No. 50-002-49311 Supplier Medchemexpress LLC Supplier No. HY10353A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Raltegravir potassium is the potassium salt of raltegravir, a potent HIV integrase strand transfer inhibitor widely used in research to study viral integration and screening assays. It is supplied in solid and solution formats and is intended for laboratory research use only.

  • Potent HIV integrase inhibitor for research applications.
  • Potassium salt form to improve handling and solubility properties.
  • Molecular formula C20H20FKN6O5, molecular weight 482.51.
  • High purity (approximately 99.8%), suitable for analytical work.
  • Available in multiple formats including 25 mg solid and 10 mM in DMSO solutions.
  • Intended for in vitro and analytical research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.