Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Raltegravir potassium | 871038-72-1 | MFCD12031642 | 99.8% | 482.51 g/mol | C20H20FKN6O5 | 1 ML
SDP

Catalog No. 5000246168
Encompass_Preferred

Raltegravir potassium is the monopotassium salt of raltegravir, an HIV-1 integrase inhibitor used in antiviral research. It is supplied as a ready-to-use 10 mM solution in DMSO and as solid quantities for formulation and assay work. Store sealed solids at 4°C; store solutions frozen for long-term stability.

  • Potassium salt of raltegravir
  • Available as 10 mM solution in DMSO and as solids
  • Molecular weight 482.51 g/mol
  • Chemical formula C20H20FKN6O5
  • High purity (≈99.8%)
  • Suitable for biochemical and cell-based assays
  • Storage: solids 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-46168 Supplier Medchemexpress LLC Supplier No. HY10353A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Raltegravir potassium is the monopotassium salt of raltegravir, an HIV-1 integrase inhibitor used in antiviral research. It is supplied as a ready-to-use 10 mM solution in DMSO and as solid quantities for formulation and assay work. Store sealed solids at 4°C; store solutions frozen for long-term stability.

  • Potassium salt of raltegravir
  • Available as 10 mM solution in DMSO and as solids
  • Molecular weight 482.51 g/mol
  • Chemical formula C20H20FKN6O5
  • High purity (≈99.8%)
  • Suitable for biochemical and cell-based assays
  • Storage: solids 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.