Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Raltegravir potassium | 871038-72-1 | MFCD12031642 | 99.8% | 482.51 g/mol | C20H20FKN6O5 | 10 MG
SDP

Catalog No. 5000248649
Encompass_Preferred

Raltegravir potassium is the potassium salt form of raltegravir, an HIV-1 integrase strand transfer inhibitor provided for research use. The compound is characterized by a defined molecular formula and high purity, with documented solubility and storage recommendations for in vitro and in vivo applications.

  • Potassium salt of raltegravir
  • HIV-1 integrase inhibitor
  • CAS number 871038-72-1
  • Molecular formula C20H20FKN6O5
  • Molecular weight 482.51 g/mol
  • Reported purity approximately 99.8%
  • Solubility: H2O 25 mg/mL; DMSO 20.83 mg/mL
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month)
  • For research use only

Catalog No. 50-002-48649 Supplier Medchemexpress LLC Supplier No. HY10353A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Raltegravir potassium is the potassium salt form of raltegravir, an HIV-1 integrase strand transfer inhibitor provided for research use. The compound is characterized by a defined molecular formula and high purity, with documented solubility and storage recommendations for in vitro and in vivo applications.

  • Potassium salt of raltegravir
  • HIV-1 integrase inhibitor
  • CAS number 871038-72-1
  • Molecular formula C20H20FKN6O5
  • Molecular weight 482.51 g/mol
  • Reported purity approximately 99.8%
  • Solubility: H2O 25 mg/mL; DMSO 20.83 mg/mL
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.