Learn More
Apexbio Technology LLC RALTEGRAVIR-POTASSIUM-SALT-5MG
SDP
Catalog No. 5000570646
Encompass_Preferred
Raltegravir potassium salt (CAS No 871038-72-1) is a potent HIV-1 integrase inhibitor extensively investigated in biomedical research Originating as an antiretroviral agent it disrupts the integration of viral DNA into the host genome a crucial step in the viral replication cycle Its mechanism of action targets the integrase enzyme effectively blocking the strand transfer reaction essential for viral replication In vitro studies often demonstrate activity in the nanomolar range making it a vital tool in antiviral research It is commonly used in various cell models including human T lymphocytes and in vivo studies to evaluate efficacy and resistance Additionally it plays a significant role in drug screening processes for novel antiretrovirals Experimental concentrations typically depend on the specific research design ensuring relevance to the study context
Catalog No. 50-005-70646 Supplier Apexbio Technology LLC Supplier No. B47605
Raltegravir potassium salt (CAS No 871038-72-1) is a potent HIV-1 integrase inhibitor extensively investigated in biomedical research Originating as an antiretroviral agent it disrupts the integration of viral DNA into the host genome a crucial step in the viral replication cycle Its mechanism of action targets the integrase enzyme effectively blocking the strand transfer reaction essential for viral replication In vitro studies often demonstrate activity in the nanomolar range making it a vital tool in antiviral research It is commonly used in various cell models including human T lymphocytes and in vivo studies to evaluate efficacy and resistance Additionally it plays a significant role in drug screening processes for novel antiretrovirals Experimental concentrations typically depend on the specific research design ensuring relevance to the study context