Learn More
Cayman Chemical Raltitrexed, Purity: ≥98%, CAS: 112887-68-0, 100 mg
SDP
Catalog No. 502936323
Encompass_Preferred
An inhibitor of thymidylate synthase (Ki = 62 nM); inhibits the growth of L1210 mouse lymphocytic leukemia cells in vitro (IC50 = 8 nM); reduces thymidylate synthase activity and completely eliminates tumors in a mouse L1210:ICR ascitic tumor model at 4 mg/kg; reduces tumor growth and the number of metastases in an HCT116-luc mouse xenograft model
Catalog No. 50-293-6323 Supplier Cayman Chemical Supplier No. 26079100 MG
An inhibitor of thymidylate synthase (Ki = 62 nM); inhibits the growth of L1210 mouse lymphocytic leukemia cells in vitro (IC50 = 8 nM); reduces thymidylate synthase activity and completely eliminates tumors in a mouse L1210:ICR ascitic tumor model at 4 mg/kg; reduces tumor growth and the number of metastases in an HCT116-luc mouse xenograft model