Learn More
Apexbio Technology LLC Raltitrexed, 112887-68-0, MFCD00864168, 10 mg
SDP
Supplier:  Apexbio Technology LLC B147610
Encompass_Preferred
MF: C21H22N4O6S, MW: 458.49, Solubility: 154mg/mL in DMSO. Raltitrexed (Tomudex, TDX, ZD 1694) is an antimetabolite drug used in cancer chemotherapy. It is an inhibitor of thymidylate synthase.
Catalog No. 50-101-1332