Learn More
Apexbio Technology LLC Raltitrexed 112887-68-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062021
Encompass_Preferred
Raltitrexed (CAS 112887-68-0) also known as Tomudex or ZD 1694 is a small-molecule antimetabolite utilized primarily in oncology research As a thymidylate synthase inhibitor raltitrexed disrupts DNA synthesis by impeding nucleotide biosynthesis thereby exerting cytotoxic effects against rapidly dividing tumor cells The compound serves as a tool in cancer studies aimed at elucidating thymidylate synthase s role in tumor cell proliferation and evaluating antitumor therapeutic approaches targeting this enzymatic pathway
Catalog No. 50-000-62021 Supplier Apexbio Technology LLC Supplier No. B14765.1
Raltitrexed (CAS 112887-68-0) also known as Tomudex or ZD 1694 is a small-molecule antimetabolite utilized primarily in oncology research As a thymidylate synthase inhibitor raltitrexed disrupts DNA synthesis by impeding nucleotide biosynthesis thereby exerting cytotoxic effects against rapidly dividing tumor cells The compound serves as a tool in cancer studies aimed at elucidating thymidylate synthase s role in tumor cell proliferation and evaluating antitumor therapeutic approaches targeting this enzymatic pathway