Learn More
Aviva Systems Biology Corp RALYL Antibody - C-terminal region (ARP41176_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41176_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RALYL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RALYL. Alternate names: HNRPCL3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-57750