Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: NA Fluoride

Catalog No. 1491570
Change view
Click to view available options
Quantity:
500/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-915-70 500/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-915-70 Supplier RAM Scientific Supplier No. 07 7340

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Pre-assembled capillary blood collection device with a capillary tube, attached pierceable cap, and a microtube

Accessory for T-Fill Capillary Blood Collection Systems

Blood samples are drawn into the SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes through capillary action, reducing hemolysis, tissue contamination, and re-draws. No scooping or milking required. Coated with Sodium Fluoride for glucose determinations.

  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No “scooping” or “milking” required
  • Capillary straw and microtube are coated with anticoagulant Lithium Heparin (Green cap) to prevent clotting
  • Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
  • Lot Number, Expiration Date, and fill lines are printed directly on the microtube
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • For plasma determinations in chemistry
  • Small sample sizes, great for pediatrics and neonates
  • Optional gel separator serves as a barrier between blood and serum after centrifugation
  • Available in a variety of sizes and draw volumes, with or without gel separator
  • Optional amber microtube ensures light-sensitive sample integrity providing protection for testing (ex. Bilirubin)

Recommended Storage

Store at room temperature. Keep dry. Keep away from sunlight.

Specifications

Color Clear
Product Type Chemistry/Glucose Tube
Additive Sodium Fluoride
Material Plastic
Sterility Non-sterile
Closure Type Standard Pierceable Cap
Capacity (Metric) 200 μL
For Use With (Equipment) T-Fill Capillary Blood Collection Systems
Closure Included Yes
Closure Color Gray
Quantity 500/Cs.
For Use With (Application) Intended for glucose and chemistry determinations.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.