Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: Tube Extender

Catalog No. 1491571
Change view
Click to view available options
Quantity:
2000/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-915-71 2000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-915-71 Supplier RAM Scientific Supplier No. 09 1004

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Intended to facilitate the transport and processing of blood samples

100% plastic Tube Extender for SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes facilitates the transport and processing of blood samples.

 

  • Pierceable attached cap with self-sealing versions
  • Anticoagulant coated capillary and tube to prevent clotting
    • Ø 13mm (width) x 95mm (length)
  • Expiration date and lot number on tube for quality control

 

Specifications

Color Clear
Product Type Tube Extender
Material Plastic
Sterility Non-sterile
For Use With (Equipment) T-Fill Capillary Blood Collection Systems
Quantity 2000/Cs.
For Use With (Application) Intended to facilitate the transport and processing of blood samples.
Description Blood Collection Tube Accessory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.