Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAM Scientific Safe-T-Fill™ Plastic Hematocrit Capillaries

Catalog No. p-2715441
Change view
Click to view available options
Additive:
Heparin
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Additive
14-915-81 None
14-915-82 Heparin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-915-81 Supplier RAM Scientific Supplier No. 06 0508
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

100% plastic—no glass to break

Blood samples are drawn into the SAFE-T-FILL™ Plastic Microhematocrit Capillary Tubes through the same capillary action as glass without the risk of breakage.

  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Replaces glass Natelson, Caraway, or mylar-coated glass microhematocrit capillary tubes
  • Blood samples are drawn into the capillary tube through the same capillary action as glass without the risk of breakage
  • Microhematocrit capillary tubes are available untreated or heparinized
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • Color-coded tip red/blue marking to designate whether additive is present in the capillary tube
  • Packaging labels are marked with Lot Number and Expiration Date

Recommended Storage

Store at room temperature. Keep dry. Keep away from sunlight.

Specifications

Color Clear with Red Mark
Product Type Microhematocrit Untreated Capillary Tube
Additive None
Material Plastic
Capacity (Metric) 50 μL
Closure Included No
For Use With (Application) Intended for microhematocrit determinations (volume of red blood cells/PCV).
Length (Metric) 7.5 cm
Length (English) 2.95 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.