Learn More
Apexbio Technology LLC Ramage Linker,Fmoc-Suberol 212783-75-0 100g
SDP
Catalog No. 5000097252
Encompass_Preferred
Ramage LinkerFmoc-Suberol is a synthetic crosslinking reagent used in peptide synthesis and protein conjugation experiments Structurally it features an Fmoc-protected functional group and a suberic acid-derived spacer arm that enables the formation of stable amide linkages with target molecules Ramage LinkerFmoc-Suberol is widely employed in antibody-drug conjugate research attachment of carrier proteins for vaccines and studies investigating protein-protein and protein-peptide interactions Based on these chemical properties Ramage LinkerFmoc-Suberol holds research potential in the development of peptide-based therapeutic agents and biomaterials
Catalog No. 50-000-97252 Supplier Apexbio Technology LLC Supplier No. A7043100000
Ramage LinkerFmoc-Suberol is a synthetic crosslinking reagent used in peptide synthesis and protein conjugation experiments Structurally it features an Fmoc-protected functional group and a suberic acid-derived spacer arm that enables the formation of stable amide linkages with target molecules Ramage LinkerFmoc-Suberol is widely employed in antibody-drug conjugate research attachment of carrier proteins for vaccines and studies investigating protein-protein and protein-peptide interactions Based on these chemical properties Ramage LinkerFmoc-Suberol holds research potential in the development of peptide-based therapeutic agents and biomaterials