Learn More
Medchemexpress LLC HY-B0745 100mg , Ramatroban BAY u3405 CAS:116649-85-5 Purity:98%
SDP
Catalog No. 501493069
Encompass_Preferred
Medchemexpress, HY-B0745 100mg Ramatroban BAY u3405 CAS:116649-85-5 Formula:C21H21FN2O4S IC50: 14 nM (hTP, TxA 2 ), 113 nM (hDP2, CRTH2), 33.4 μM (hDP1), 15 μM (CYP2C9) Purity:98% Ramatroban is a selective thromboxane A 2 ( TxA 2 , IC 50 =14 nM) antagonist, which also antagonizes CRTH2 ( IC 50 =113 nM) by inhibiting PGD 2 binding. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3069 Supplier Medchemexpress LLC Supplier No. HYB0745100MG
Medchemexpress, HY-B0745 100mg Ramatroban BAY u3405 CAS:116649-85-5 Formula:C21H21FN2O4S IC50: 14 nM (hTP, TxA 2 ), 113 nM (hDP2, CRTH2), 33.4 μM (hDP1), 15 μM (CYP2C9) Purity:98% Ramatroban is a selective thromboxane A 2 ( TxA 2 , IC 50 =14 nM) antagonist, which also antagonizes CRTH2 ( IC 50 =113 nM) by inhibiting PGD 2 binding. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.