Learn More
TARGETMOL CHEMICALS INC RAMATROBAN 50MG
SDP
Catalog No. 502580838
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ramatroban (BAY u3405) (BAY u3405) is a thromboxane A(2) (TxA(2)) antagonist marketed for allergic rhinitis. It has been used in trials studying the treatment of Asthma. purity: 99%
Catalog No. 50-258-0838 Supplier TARGETMOL CHEMICALS INC Supplier No. T239650MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ramatroban (BAY u3405) (BAY u3405) is a thromboxane A(2) (TxA(2)) antagonist marketed for allergic rhinitis. It has been used in trials studying the treatment of Asthma. purity: 99%