Learn More
Medchemexpress LLC Ramelteon-d3 | 1432057-38-9 | 99.0% | C16H18D3NO2 | 5 MG
SDP
Catalog No. 5000433305
Encompass_Preferred
Ramelteon-d3 is a deuterium-labelled Ramelteon, functioning as a potent and highly selective MT1/MT2 agonist. It is used in insomnia research, consistently reducing sleep onset without withdrawal symptoms. This compound serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potent and highly selective MT1/MT2 agonist
  • Reduces sleep onset in insomnia research
  • No next-morning residual effects or rebound insomnia
  • No withdrawal symptoms upon discontinuation
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for analytical methods
Catalog No. 50-004-33305 Supplier Medchemexpress LLC Supplier No. HYA0014S15MG
Add to Cart
Add to Cart
Ramelteon-d3 is a deuterium-labelled Ramelteon, functioning as a potent and highly selective MT1/MT2 agonist. It is used in insomnia research, consistently reducing sleep onset without withdrawal symptoms. This compound serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potent and highly selective MT1/MT2 agonist
  • Reduces sleep onset in insomnia research
  • No next-morning residual effects or rebound insomnia
  • No withdrawal symptoms upon discontinuation
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for analytical methods