Learn More
Medchemexpress LLC Ramelteon metabolite M-II-d3 | 1246812-22-5 | 99.8% | 278.36 | C16H18D3NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY103005S1MG
Encompass_Preferred
Ramelteon metabolite M-II-d3 is the deuterium-labeled form of Ramelteon metabolite M-II, provided for use as an internal standard or tracer in quantitative analytical methods. It is intended for analytical and research applications where a labeled reference compound improves accuracy and precision in mass spectrometry and NMR workflows.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High reported purity for accurate quantitation (≈99.8%).
  • Provided in milligram-scale pack sizes for analytical use.
  • Molecular weight 278.36 and chemical formula C16H18D3NO3.
Catalog No. 50-002-48445