Learn More
Medchemexpress LLC Ramelteon metabolite M-II-d3 | 1246812-22-5 | 99.8% | 278.36 g/mol | C16H18D3NO3 | 5 MG
SDP
Catalog No. 5000245483
Encompass_Preferred
Ramelteon metabolite M-II-d3 is the deuterium-labeled analog of the major human metabolite of ramelteon, provided as a high-purity reference standard intended for use as a tracer or internal standard in quantitative analytical methods. It is supplied as a solid suitable for laboratory analysis.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity suitable for analytical work.
  • Supplied as a white to off-white solid for ease of handling.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Molecular weight approximately 278.36 g/mol for accurate calculations.
Catalog No. 50-002-45483 Supplier Medchemexpress LLC Supplier No. HY103005S5MG
Ramelteon metabolite M-II-d3 is the deuterium-labeled analog of the major human metabolite of ramelteon, provided as a high-purity reference standard intended for use as a tracer or internal standard in quantitative analytical methods. It is supplied as a solid suitable for laboratory analysis.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity suitable for analytical work.
  • Supplied as a white to off-white solid for ease of handling.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Molecular weight approximately 278.36 g/mol for accurate calculations.