Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MarqMetrix All-In-One Process Raman Analyzer

Catalog No. 18001153
Change view
Click to view available options
Type:
Process Raman Analyzer
Includes:
Bioreactor Ball Probe
Low-Pressure Flow Cell
Process Ball Probe
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Includes
18-001-153 Process Raman Analyzer Low-Pressure Flow Cell
18-001-151 Process Raman Analyzer Bioreactor Ball Probe
18-001-152 Process Raman Analyzer Process Ball Probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-001-153 Supplier Thermo Scientific™ Supplier No. 912A1262
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Thermo Scientific™ MarqMetrix™ All-In-One Process Raman Analyzer is a compact and portable Raman spectrometer all-in-one system with patented technology, purpose-built for rapid deployment, ease-of-use, and flexible scalability in markets where time-to-results is key.

With the MarqMetrix Process Raman Analzyer, businesses can conduct real-time, non-destructive, and immediate analysis without sample preparation.

The MarqMetrix All-In-One analyzer makes precise and accurate measurements simple to execute and understand. Our one-button hardware and easy-to-use software will have you taking highly accurate Raman measurements in less than 15 minutes out of the box.

The MarqMetrix All-In-One System is easily integrated into your existing process and eliminates the need for costly technical expertise.

Application versatility

  • Bioprocessing PAT (Process Analytical Technology)
  • Oil & Gas production

Use Raman inline for process monitoring

With a small footprint and lower cost, the MarqMetrix All-In-One Process Raman Analyzer makes process monitoring possible in-line and enables you to make optimization decisions faster.

Pack your MarqMetrix All-In-One System in a protective case and take it to the point of need. Factory calibration ensures continuous and precise analysis out of the box.

Service Plans

Thermo Fisher Scientific offers Service Plans designed to maximize productivity and optimize performance throughout the lifecycle of your MarqMetrix All-In-One system. Plans include exclusive service advantages such as:

  • Unlimited technical support
  • Depot repair services
  • Maintaining system compliance
  • Consulting services

Software

Maximize the power of the MarqMetrix All-In-One Process Raman Analyzer with Thermo Scientific Lykos PAT Software - a remote control and monitoring software designed to simplify the complexities of chemometrics with features including:

  • Real-time display, storage, and export of bioprocess monitoring results
  • Remotely control and monitor the MarqMetrix All-In-One Process Raman Analyzer from outside of the lab
  • Simplified use and intuitive workflow for non-Ramen spectroscoptists
  • Control multiple MarqMetrix All-In-One analyzers from a central location

Available add-ons: cGMP and 21 CFR part 11 compliance features for regulated environments

Specifications

Type Process Raman Analyzer
Depth (Metric) 25 cm
Electrical Requirements Input voltage: 90-264 VAC, 50-60 Hz, Operating voltage: 12VDC, 2A
Height (Metric) 7.7 cm
Resolution 6.5 cm-1 average across spectral range
Software Details Software: MarqMetrix Raman suiteController: Integrated PC (Windows 10 OS)
Spectral Range 100 to 3250 cm-1
Wavelength Range 785 nm, Excitation power: 10 to 450 mW
Width (Metric) 25 cm
Safety Features 1 keyed Laser interlock switch, Electronic laser trigger
Detector Thermally stabilized CCD (2 stage TEC)
Fiber Conditions FC/PC (Both excitation and collection)
Includes Low-Pressure Flow Cell
Certifications/Compliance Laser Certification: Class 3B laser deviceReported to the FDA
Dark Noise ≤16.5 counts at 10 sec. integration
Ports 1RJ-45 Ethernet port 3 USB 3.0, 1 Display port 1.1 (VGA, DVI or HDMI with adaptor)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.