Learn More
Thermo Scientific™ MarqMetrix Replacement FlowCell BallProbe
Catalog No. 18001157
Change view
Click to view available options
Type:
Replacement FlowCell BallProbe
Catalog No. Type
18-001-157 Replacement FlowCell BallProbe
1 options
This item is not returnable. View return policy
Catalog No. 18-001-157 Supplier Thermo Scientific™ Supplier No. 840402700
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Replacement BallProbe for use with the MarqMetrix All-In-One process raman analyzer.
  • Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
Specifications
For Use With (Equipment) MarqMetrix All-In-One Process Raman Analyzer
Type Replacement FlowCell BallProbe