Learn More
Medchemexpress LLC Ramipril-d5 (HOE-498-d5) | 1132661-86-9 | 99.0% | 1 MG
SDP
Catalog No. 5000406002
Encompass_Preferred
Ramipril-d5 is a deuterium-labeled form of Ramipril, a potent angiotensin-converting enzyme (ACE) inhibitor with an IC50 of 5 nM. It is specifically designed for research applications, serving as a reliable tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes like deuterium plays a crucial role in drug development, particularly for accurate quantitation, and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled form of Ramipril
  • Potent ACE inhibitor with IC50 of 5 nM
  • Suitable for research applications
  • Functions as a tracer and internal standard
  • Compatible with quantitative analysis methods (NMR, GC-MS, LC-MS)
  • Supports drug development by aiding in quantitation
  • May influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06002 Supplier Medchemexpress LLC Supplier No. HYB0279S1MG
Ramipril-d5 is a deuterium-labeled form of Ramipril, a potent angiotensin-converting enzyme (ACE) inhibitor with an IC50 of 5 nM. It is specifically designed for research applications, serving as a reliable tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes like deuterium plays a crucial role in drug development, particularly for accurate quantitation, and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled form of Ramipril
  • Potent ACE inhibitor with IC50 of 5 nM
  • Suitable for research applications
  • Functions as a tracer and internal standard
  • Compatible with quantitative analysis methods (NMR, GC-MS, LC-MS)
  • Supports drug development by aiding in quantitation
  • May influence pharmacokinetic and metabolic profiles of drugs