Learn More
Cayman Chemical RamIprIl DIketopIperazIn 25mg
SDP
Catalog No. 502910920
Encompass_Preferred
A potential impurity found in commercial preparations of ramipril
Catalog No. 50-291-0920 Supplier Cayman Chemical Supplier No. 3491725 MG
A potential impurity found in commercial preparations of ramipril