Learn More
Medchemexpress LLC Ramipril (HOE-498) | 87333-19-5 | 99.4% | 1 ML
SDP
Catalog No. 5000409017
Encompass_Preferred
Ramipril (HOE-498) is an angiotensin-converting enzyme (ACE) inhibitor with an IC50 of 5 nM. It enhances the activity of ACE-associated CK2 and the phosphorylation of ACE Ser1270 in cultured endothelial cells. Chronic in vivo administration to rats significantly reduces LPS-induced apoptosis. It inhibits systolic blood pressure (SBP) with an IC50 of 1.97 mg/kg in spontaneously hypertensive rats (SHR).
  • Potent angiotensin-converting enzyme (ACE) inhibitor
  • Enhances ACE-associated CK2 activity
  • Reduces LPS-induced apoptosis in rats
  • Inhibits systolic blood pressure (SBP)
  • For research use only
Catalog No. 50-004-09017 Supplier Medchemexpress LLC Supplier No. HYB027910MM/1ML
Ramipril (HOE-498) is an angiotensin-converting enzyme (ACE) inhibitor with an IC50 of 5 nM. It enhances the activity of ACE-associated CK2 and the phosphorylation of ACE Ser1270 in cultured endothelial cells. Chronic in vivo administration to rats significantly reduces LPS-induced apoptosis. It inhibits systolic blood pressure (SBP) with an IC50 of 1.97 mg/kg in spontaneously hypertensive rats (SHR).
  • Potent angiotensin-converting enzyme (ACE) inhibitor
  • Enhances ACE-associated CK2 activity
  • Reduces LPS-induced apoptosis in rats
  • Inhibits systolic blood pressure (SBP)
  • For research use only