Learn More
AdipoGen Ramoplanin A2 (5 mg)
SDP
Supplier:  AdipoGen AGCN20318M005
Encompass_Preferred
Ramoplanin A2 (5 mg), Adipogen Life Sciences. Chemical. CAS: 81988-88-7 [A2]. Formula: C119H154ClN21O40. MW: 2554.1. Isolated from Actinoplanes sp. Glycolipodepsipeptide antibiotic. Complex of structurally related molecules A1, A2 and A3, with ramoplanin A2 as the primary component. Antibacterial and antiviral agent with activity against aerobic and anaerobic Gram-positive bacteria such as Clostridium difficile and antibiotic-resistant enterococci. Inhibits cell wall synthesis and consequently bacterial growth by forming a complex with lipid intermediate II (Lipid II), a key intermediate in peptidoglycan biosynthesis.
Catalog No. 50-168-7326