Learn More
Cayman Chemical RamoplnIn Complex 100mg
SDP
Catalog No. 502940870
Encompass_Preferred
An antibiotic complex; active against clinical isolates of S. aureus, S. pyogenes, S. pneumoniae, and S. faecalis (MICs = 0.016-2 UG/ml) but not Gram-negative bacteria or C. albicans (MICs = ≥128 UG/ml); inhibits cell wall synthesis in B. subtilis at 0.5 UG/ml; effective against systemic S. pyogenes, S. pneumoniae, and S. aureus infections in mice (ED50s = 0.081, 0.14, and ~25 mg/kg per day, respectively)
Catalog No. 50-294-0870 Supplier Cayman Chemical Supplier No. 28189100 MG
An antibiotic complex; active against clinical isolates of S. aureus, S. pyogenes, S. pneumoniae, and S. faecalis (MICs = 0.016-2 UG/ml) but not Gram-negative bacteria or C. albicans (MICs = ≥128 UG/ml); inhibits cell wall synthesis in B. subtilis at 0.5 UG/ml; effective against systemic S. pyogenes, S. pneumoniae, and S. aureus infections in mice (ED50s = 0.081, 0.14, and ~25 mg/kg per day, respectively)