Learn More
Cayman Chemical Ramoplanin Complex, Purity: ≥80% (Ramoplanin A2), CAS: 76168-82-6, 250 mg
SDP
Catalog No. 502938609
Encompass_Preferred
An antibiotic complex; active against clinical isolates of S. aureus, S. pyogenes, S. pneumoniae, and S. faecalis (MICs = 0.016-2 µg/ml) but not Gram-negative bacteria or C. albicans (MICs = ≥128 µg/ml); inhibits cell wall synthesis in B. subtilis at 0.5 µg/ml; effective against systemic S. pyogenes, S. pneumoniae, and S. aureus infections in mice (ED50s = 0.081, 0.14, and ~25 mg/kg per day, respectively)
Catalog No. 50-293-8609 Supplier Cayman Chemical Supplier No. 28189250 MG
An antibiotic complex; active against clinical isolates of S. aureus, S. pyogenes, S. pneumoniae, and S. faecalis (MICs = 0.016-2 µg/ml) but not Gram-negative bacteria or C. albicans (MICs = ≥128 µg/ml); inhibits cell wall synthesis in B. subtilis at 0.5 µg/ml; effective against systemic S. pyogenes, S. pneumoniae, and S. aureus infections in mice (ED50s = 0.081, 0.14, and ~25 mg/kg per day, respectively)