Learn More
ATCC Ramos (RA 1); Burkitt's Lymphoma; Human (Homo sapiens)
Catalog No. 502383949
Encompass
Ramos (RA 1) is a B lymphocyte cell line that was derived from a 3-year-old, White, male patient with Burkitt’s Lymphoma (American). This cell line can be used in immunology research.
Catalog No. 50-238-3949 Supplier ATCC Supplier No. CRL1596
Ramos (RA 1) is a B lymphocyte cell line that was derived from a 3-year-old, White, male patient with Burkitt’s Lymphoma (American). This cell line can be used in immunology research.