Learn More
Medchemexpress LLC Ramosetron (Hydrochloride) | 132907-72-3 | 100.0% | 1 ML
SDP
Catalog No. 5000408388
Encompass_Preferred
Ramosetron Hydrochloride is a serotonin 5-HT3 receptor antagonist, primarily investigated for the treatment of nausea and vomiting, as well as irritable bowel syndrome with diarrhea. It exhibits potent and long-acting effects with high selectivity and affinity for its target.
  • Serotonin 5-HT3 receptor antagonist
  • Investigated for irritable bowel syndrome with diarrhea
  • Effective on chemotherapy-induced nausea and vomiting (CINV)
  • Effective on postoperative nausea and vomiting (PONV)
  • Potent and long-acting effects
  • High selectivity and affinity for 5-HT3 receptors
Catalog No. 50-004-08388 Supplier Medchemexpress LLC Supplier No. HYB059510MM/1ML
Ramosetron Hydrochloride is a serotonin 5-HT3 receptor antagonist, primarily investigated for the treatment of nausea and vomiting, as well as irritable bowel syndrome with diarrhea. It exhibits potent and long-acting effects with high selectivity and affinity for its target.
  • Serotonin 5-HT3 receptor antagonist
  • Investigated for irritable bowel syndrome with diarrhea
  • Effective on chemotherapy-induced nausea and vomiting (CINV)
  • Effective on postoperative nausea and vomiting (PONV)
  • Potent and long-acting effects
  • High selectivity and affinity for 5-HT3 receptors