Learn More
Medchemexpress LLC Ramosetron Hydrochloride | 132907-72-3 | 99.95% | 100 MG
SDP
Catalog No. 5000409087
Encompass_Preferred
Ramosetron Hydrochloride (YM060) is an orally active 5-HT3 antagonist. It is promising for research into irritable bowel syndrome with diarrhea, improving dysphoria, interference with activity, and food avoidance. It ameliorates 5-fluorouracil (5-FU)-induced intestinal mucositis in C57BL/6 mice.
  • Orally active 5-HT3 antagonist
  • Improves dysphoria and food avoidance
  • Promising for irritable bowel syndrome with diarrhea research
  • Ameliorates 5-FU-induced intestinal mucositis
  • Targets the 5-HT3 receptor
Catalog No. 50-004-09087 Supplier Medchemexpress LLC Supplier No. HYB0595100MG
Ramosetron Hydrochloride (YM060) is an orally active 5-HT3 antagonist. It is promising for research into irritable bowel syndrome with diarrhea, improving dysphoria, interference with activity, and food avoidance. It ameliorates 5-fluorouracil (5-FU)-induced intestinal mucositis in C57BL/6 mice.
  • Orally active 5-HT3 antagonist
  • Improves dysphoria and food avoidance
  • Promising for irritable bowel syndrome with diarrhea research
  • Ameliorates 5-FU-induced intestinal mucositis
  • Targets the 5-HT3 receptor