Learn More
Selleck Chemical LLC Ramosetron Hydrochloride S3723-25mg
SDP
Catalog No. 503108947
Encompass_Preferred
Ramosetron Hydrochloride (YM-060) is the hydrochloride salt of ramosetron a selective serotonin (5-HT) receptor antagonist with potential antiemetic activity
Catalog No. 50-310-8947 Supplier Selleck Chemical LLC Supplier No. S372325MG
Ramosetron Hydrochloride (YM-060) is the hydrochloride salt of ramosetron a selective serotonin (5-HT) receptor antagonist with potential antiemetic activity