Learn More
Cayman Chemical Ramosetron (hydrochloride), Purity: ≥98%, CAS: 132907-72-3, 10 mg
SDP
Catalog No. 502969811
Encompass_Preferred
A potent and selective antagonist of the serotonin (5-HT) receptor subtype 5-HT3 (Ki = 0.06 nM); has little or no effect at other 5-HT receptor subtypes or at adrenergic or histamine receptors
Catalog No. 50-296-9811 Supplier Cayman Chemical Supplier No. 1554810 MG
A potent and selective antagonist of the serotonin (5-HT) receptor subtype 5-HT3 (Ki = 0.06 nM); has little or no effect at other 5-HT receptor subtypes or at adrenergic or histamine receptors