Learn More
Montana Molecular RAMP3 Expression Kit - Big Sky BacMam, 5mL - Montana Molecular
SDP
Catalog No. NC3787570
Encompass_Preferred
Receptor Activity Modifying Protein 3 (RAMP3) expression vector - packaged in Montana Molecular's Big Sky BacMam for improved expression in mammalian cells across cell lines, primary cultures, and iPSCs. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity, enabling rapid assay optimization. Co-express with other Montana Molecular tools including the Calcitonin receptor-like receptor and cAMP or Arrestin recruitment biosensors.
Catalog No. NC3787570 Supplier Montana Molecular Supplier No. R0003N005
Receptor Activity Modifying Protein 3 (RAMP3) expression vector - packaged in Montana Molecular's Big Sky BacMam for improved expression in mammalian cells across cell lines, primary cultures, and iPSCs. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity, enabling rapid assay optimization. Co-express with other Montana Molecular tools including the Calcitonin receptor-like receptor and cAMP or Arrestin recruitment biosensors.