Learn More
Medchemexpress LLC HY-P9920 5mg , Ramucirumab CAS:947687-13-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99205MG
Encompass_Preferred
HY-P9920 5mg Ramucirumab CAS: 947687-13-0 Ramucirumab is a human VEGFR-2 antagonist for the treatment of solid tumors. Ramucirumab is a recombinant human immunoglobulin G1 monoclonal antibody that binds to the extracellular binding domain of VEGFR-2 and prevents the binding of VEGFR ligands: VEGF-A, VEGF-C, and VEGF-D. Ramucirumab is also an angiogenesis inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9847