Learn More
Aviva Systems Biology Corp RAN Antibody - middle region (ARP56714_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56714_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RAN. Alternate names: ARA24, Gsp1, TC4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-61961