Learn More
Aviva Systems Biology Corp RANBP1 Antibody - C-terminal region (ARP46083_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46083_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RANBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RANBP1. Alternate names: MGC88701, HTF9AApplicationWB. Species reactivity: Cow, Horse, Human, Yeast
Catalog No. 50-014-59216