Learn More
Aviva Systems Biology Corp RANBP3 Antibody - N-terminal region (ARP58520_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58520_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RANBP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RANBP3. Alternate names: DKFZp586I1520ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-62339