Learn More
Aviva Systems Biology Corp RANBP5 Antibody - N-terminal region (ARP53600_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53600_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RANBP5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RANBP5. Alternate names: DKFZp686O1576, FLJ43041, IMB3, KPNB3, MGC2068, RANBP5, imp5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-856