Learn More
World Precision Instrument Randall Selitto Paw Pressure Meter, 800g
SDP
Catalog No. 502538778
Encompass
Get instantaneous, live readings
The Randall Selitto Paw Pressure Meter for analgesia testing is digitally controlled. Use the handheld instrument to apply force to an animal’s extremity and get instantaneous, live readings. You can even view the last maximum force applied during the test. The new limit indicator lets you select the maximum force limit, and then indicates with a warning light when the system reaches that limit. This unit comes with an acrylic stand to allow for easy viewing of the display. Stand and sling are sold separately.
Catalog No. 50-253-8778 Supplier World Precision Instrument Supplier No. II2500
Add to Cart
Add to Cart
Request a Quote
Get instantaneous, live readings
The Randall Selitto Paw Pressure Meter for analgesia testing is digitally controlled. Use the handheld instrument to apply force to an animal’s extremity and get instantaneous, live readings. You can even view the last maximum force applied during the test. The new limit indicator lets you select the maximum force limit, and then indicates with a warning light when the system reaches that limit. This unit comes with an acrylic stand to allow for easy viewing of the display. Stand and sling are sold separately.