Learn More
Takara Bio RANDOM PRIM LABELING KIT 30RXN
SDP
Catalog No. 50443849
Encompass
DNA labeling kit; Random primer; Version 2; 30 reactions; Used for labeling DNA for use in hybridization probes
Catalog No. 50-443-849 Supplier Takara Bio Supplier No. 6045
DNA labeling kit; Random primer; Version 2; 30 reactions; Used for labeling DNA for use in hybridization probes