Learn More
New England Biolabs, Inc. Random Primer Mix – 100 µl
SDP
Catalog No. 504484
Encompass
It is recommended to use the Random Primer Mix for reverse transcription of the following RNA templates: 1) RNA without poly(A) tail such as ribosomal RNAs 2) RNA with strong secondary structures 3) Partially degraded RNA samples 4) Target regions at 5' end of a long messenger RNA transcript
Catalog No. 50-448-4 Supplier New England Biolabs, Inc. Supplier No. S1330S
It is recommended to use the Random Primer Mix for reverse transcription of the following RNA templates: 1) RNA without poly(A) tail such as ribosomal RNAs 2) RNA with strong secondary structures 3) Partially degraded RNA samples 4) Target regions at 5' end of a long messenger RNA transcript