Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Raney™-Nickel

Catalog No. 510068100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-006-8100G 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 51-006-8100G Supplier Sigma Aldrich Supplier No. 510068100G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Useful as a selective reducing agent for hydrogenation of organic compoundsRaney is a registered trademark of W.R. Grace and Co.-Conn.

Specifications

Melting Point -39°C
Density 1.13 g/mL at 25°C
MDL Number MFCD00081864
Quantity 250 mL
Synonym Tetrabutyl titanate polymer; Tilcom PBT; Titanium tetrabutoxide polymer; Tytan PBT; Tyzor BTP; Tyzor PB; Vertec PBT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.