Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Raney™-Nickel

Catalog No. 221678100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-167-8100G 100 g
22-167-8500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 22-167-8100G Supplier Sigma Aldrich Supplier No. 221678100G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used as reducing agent for hydrogenation of organic compounds, e.g. hydrogenation of nitriles to amines, reduction of aromatic groups to the corresponding saturated compounds, hydrogenation reaction of beta-keto esters, rreduction of amination, and also used as 3-alkylation of oxindole with alcohol, and Ullman-type cross coupling, and dehalogenation reactions.Raney is a registered trademark of W.R. Grace and Co.-Conn.

Specifications

CAS 7440-02-0
Linear Formula AlNi
MDL Number MFCD00167392
Quantity 100 g
Synonym Raney Ni; Nickel sponge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.