Learn More
Aviva Systems Biology Corp RANGAP1 Antibody - N-terminal region (ARP56509_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56509_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RANGAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RANGAP1. Alternate names: Fug1, KIAA1835, MGC20266, SDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61886