Learn More
Honeywell™ Ranger™ Dielectric Pacs Boots
Catalog No. 19024969
Change view
Click to view available options
Size:
12
Catalog No. Size
19-024969 12
1 options
This item is not returnable. View return policy
Catalog No. 19-024969 Supplier Honeywell™ Supplier No. R6130RDM120
This item is not returnable. View return policy
Over-the-sock electrical hazard boots
  • Premium-grade rubber construction with safety yellow trim
  • Special ozone-resistant rubber compound
  • Flexible, hand-layered construction
  • Durable nylon linings for easy on/off
  • Ultra-composite shanks reinforce construction and support the feet
  • Height: 12 (30.5cm)
Not for use near explosives or where conductive footwear is required.
Compliance
Steel Toe meets ASTM F2413-11 M I/75 C/75 and EH
TRUSTED_SUSTAINABILITY
Specifications
Size 12