Learn More
Medchemexpress LLC Ranitidine | 66357-35-5 | MFCD00081180 | 99.9% | 1 ML
SDP
Catalog No. 5000407067
Encompass_Preferred
Ranitidine is a potent, selective, and orally active histamine H2-receptor antagonist. It effectively inhibits gastric secretion and antagonizes histamine-induced increases in guinea-pig isolated rat atrium and relaxations of the rat isolated uterine horn. Research indicates its potential in inhibiting breast tumor development and spread in mice, as well as reducing CD11b+Ly6Chi cells and lung metastasis.
  • Potent, selective, and orally active histamine H2-receptor antagonist
  • Inhibits gastric secretion
  • Antagonizes Histamine-induced effects in isolated heart and uterine horn
  • Inhibits breast tumor development and spread
  • Reduces CD11b+Ly6Chi cells and inhibits lung metastasis
  • Inhibits primary tumor growth
  • Increases the latency of breast tumorigenesis and reduces tumor numbers
Catalog No. 50-004-07067 Supplier Medchemexpress LLC Supplier No. HYB069310MM/1ML
Ranitidine is a potent, selective, and orally active histamine H2-receptor antagonist. It effectively inhibits gastric secretion and antagonizes histamine-induced increases in guinea-pig isolated rat atrium and relaxations of the rat isolated uterine horn. Research indicates its potential in inhibiting breast tumor development and spread in mice, as well as reducing CD11b+Ly6Chi cells and lung metastasis.
  • Potent, selective, and orally active histamine H2-receptor antagonist
  • Inhibits gastric secretion
  • Antagonizes Histamine-induced effects in isolated heart and uterine horn
  • Inhibits breast tumor development and spread
  • Reduces CD11b+Ly6Chi cells and inhibits lung metastasis
  • Inhibits primary tumor growth
  • Increases the latency of breast tumorigenesis and reduces tumor numbers