Learn More
Medchemexpress LLC Ranitidine | 66357-35-5 | MFCD00081180 | 99.9% | 25 MG
SDP
Catalog No. 5000450674
Encompass_Preferred
Ranitidine is a potent, selective, and orally active histamine H2-receptor antagonist that inhibits gastric secretion. It has been shown to inhibit breast tumor development and spread in mice, and also antagonizes histamine-induced effects in isolated rat tissues.
  • Potent, selective, and orally active H2-receptor antagonist
  • Inhibits gastric acid secretion
  • Antagonizes histamine-induced effects in isolated rat atrium and uterine horn
  • Inhibits breast tumor development and spread
  • Reduces specific myeloid cell populations in mice
  • Increases latency of breast tumorigenesis
Catalog No. 50-004-50674 Supplier Medchemexpress LLC Supplier No. 25MGHYB0693
Ranitidine is a potent, selective, and orally active histamine H2-receptor antagonist that inhibits gastric secretion. It has been shown to inhibit breast tumor development and spread in mice, and also antagonizes histamine-induced effects in isolated rat tissues.
  • Potent, selective, and orally active H2-receptor antagonist
  • Inhibits gastric acid secretion
  • Antagonizes histamine-induced effects in isolated rat atrium and uterine horn
  • Inhibits breast tumor development and spread
  • Reduces specific myeloid cell populations in mice
  • Increases latency of breast tumorigenesis